Learn More
Fisherbrand™ Chemical Absorbent Pads
Fisherbrand™ Chemical Absorbent Pads absorb chemicals and hazardous liquids
Brand:  Fisherbrand™ FBEU100E
210.97 EUR valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Product Code. 15853720
  • 297.44 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • One sided spunbound gives extra strength as well as less lint
  • Does not degrade or cause a dangerous chemical reaction with the absorbed liquid
  • Green color alerts workers that the product is being used for hazardous liquids and allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Perforated for more flexibility - use only what you need and minimize waste
  • Perfect to clean up chemical spills or for use in laboratories
Specifications
Specifications
Green
89 L
Chemical Sorbent
100 Pads/Cs.
50 x 40 cm
40 cm
15.65 lb.
Pads, 50 x 40 cm, Medium-weight, Perforated and bonded
Spunbond, Meltblown
Overspray, leaks and drips in drum and liquid storage areas, workstations and laboratories
2-ply
50 cm
16.14 in.
7.1 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Chemical Absorbent Pads > Chemical Absorbent Pads