Learn More
Thermo Scientific™ Chemical Kit for TruScan G3/RM/GP
Raman Spectrometer Certification Kit for the Analytical Instrument Qualification of TruScan, TruScan RM or TruScan GP. Kit to be used in conjunction with IQ/OQ/PQ document.
Brand:  Thermo Scientific™ 580-01501-01
Product Code. 18021081
  • 908.00 € / Each
Explore more special offers
Description
Description
  • Certification kit validity is good for one year from date of manufacture
  • Kit consists of qualified sample lots of Polystyrene, Cyclohexane, and Acetaminophen
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ramina Process Analyzer
Certification Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Chemical Kit for TruScan G3/RM/GP > Chemical Kit for TruScan G3/RM/GP; For Use With (Equipment): Ramina Process Analyzer