Learn More
Thermo Scientific™ TruScan™ G3 Handheld Raman Analyzer
Rapid material identification testing for pharmaceutical and biotechnology manufacturing quality control/quality assurance processes.
Brand: Thermo Scientific™ 800-04791-01
Description
The TruScan G3 Handheld Raman Analyzer continues the evolution of raw material ID testing for pharmaceutical and biotechnology manufacturing quality control processes requires speed, ease and efficiency to maintain continuous manufacturing. Samples are often brought to a quality control lab for testing, increasing costs and decreasing productivity. The TruScan G3 Handheld Raman Analyzer enables non-technical operators to rapidly perform raw material ID testing without the need of a lab, bringing lab-quality performance and results to any setting, whether that be a warehouse or loading dock.
- Avoid compendial ID testing that requires extensive time and laboratory training by moving ID testing to the point of receipt.
- Non-destructive, point and shoot sampling through plastic bags, glass containers, blister packs and clear gel caps.
- Backwards compatible with Truscan RM Handheld Raman Analyzer validated methods, making deploying the TruScan G3 Handheld Raman Analyzer a quick method transfer for a near seamless experience.
- A secured method for transferring of data via WIFI or wired connection for peace of mind.
- No required, frequent hardware maintenance or consumables to operates
- Enhanced 21 CFR Part 11 compliance

Specifications
Most linear and 2D standards | |
FDA 1040, 21 CFR part 11, CE certification, Ph. Eur. 8.7 | |
Ethernet, WiFi (optional) |
|
7.8 x 4.6 x 1.8 in (19.9 x 11.8 x4.6 cm) |
|
785nm ± 0.5 (2 cm-1 line width, stability <0.1 cm-1) | |
OS: Windows 10 and 11; Browers: Edge and Chrome |
|
USBc and Power |
|
250 to 2875 cm-1 | |
-20°C to 50°C |
|
0.95 kg |
Rechargeable internal lithium ion battery >3 hours operation |
|
NA = 0.33, 18 mm working distance; 0.2 mm spot size |
|
TruScan G3 Handheld Raman Analyzer | |
Exposure time set automatically |
|
250 mW ±25 | |
Vial holder, extension probe, nose cone, hot swap batteries, hand strap, tablet holder |
|
DC Wall Adapter 100/240 V, 50/60 Hz | |
8 to 10.5 cm-1(FWHM) across range | |
2.1 lb |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.