Learn More
WTW™ Chemical Oxygen Demand (COD) Test Kits
Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.
Brand: WTW™ 250351
129.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
- Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
- Safe determinations of COD in water
- Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
- For waste water, production control, ground water, surface water
- For PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, spectroFlex photometers
Specifications
EN ISO 15705 | |
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Ph | |
300 to 3500 mg/L |
Wastewater, production control, ground water, surface water | |
300 | |
CombiCheck Aluminum (Al) reagent test kit |
Safety and Handling
- COD Cell Test Method: photometric 300 - 3500 mg/l Spectroquant
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH208-Contains potassium dichromate. May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, mercury(II) sulphate, potassium dichromate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.