missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chemical Oxygen Demand (COD) Test Kits

Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.

127.00 € - 136.00 €


Products 4
Product Code Brand Range Price Quantity & Availability  
Product Code Brand Range Price Quantity & Availability  
11727315
View Documents
WTW™
252071
25 to 1500 mg/L
135.00 €
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
11774725
View Documents
WTW™
250351
300 to 3500 mg/L
136.00 €
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
11701083
View Documents
WTW™
250303
4 to 40 mg/L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11791073
View Documents
WTW™
250309
6 at 20°C
136.00 €
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
  • Safe determinations of COD in water
  • Chlorine dioxide (ClO2) reagent test kit
  • For waste water, production control, ground water, surface water
  • For PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD Cell Test Method: photometric Spectroquant

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury(II) sulphate

SDS
Documents

Documents

Product Certifications