Learn More
Thermo Scientific™ Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Brand:  Thermo Scientific™ 398184
 View more versions of this product
Product Code. 12623115
  • 138.69 € / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Rack Tower
Silver
Stainless Steel
3 cu. ft. chest freezers
14 cm
38.6 cm
5.5 in.
2 in. Box Rack - Side Access Rack (Tower)
Boxes (2 in.)
7 Boxes/Rack
3 cu. ft.
5.5 in.
15.2 in.
14 cm