Learn More
Thermo Scientific™ Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Brand:  Thermo Scientific™ 398187
 View more versions of this product
Product Code. 12633115
  • 132.66 € / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Rack Tower
Silver
Stainless Steel
13, 17, and 20 cu. ft. freezers
14 cm
67.3 cm
5.5 in.
2 in. Box Rack - Side Access Rack (Tower)
Boxes (3 in.)
8 Boxes/Rack
13, 17, 20 cu. ft.
5.5 in.
26.5 in.
14 cm