Learn More
Invitrogen™ Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647
Chicken Polyclonal Secondary Antibody
Brand: Invitrogen™ A21443
247.00 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
This chicken anti-rabbit antibody reacts with IgG heavy chains and all classes of immunoglobulin light chains from rabbit. Chicken secondary antibodies have gained popularity because they demonstrate a lower level of nonspecific binding. Chicken antibodies lack a classic Fc domain and will not bind to protein A or protein G, nor will they bind to mammalian IgG Fc receptors.Fluorescence of this long-wavelength Alexa Fluor dye is not visible by looking through a conventional fluorescence microscope. Product will be shipped at Room Temperature.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 647 | |
Chicken | |
purified | |
RUO | |
Rabbit | |
Secondary Antibody | |
IgY |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains. | |
1 mg | |
Secondary | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.