missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-3,3,3-trifluoropropylsilane, 96%, Thermo Scientific™

54.00 € - 508.00 €

Chemical Identifiers

CAS 1481-41-0
Molecular Formula C5H10ClF3Si
Molecular Weight (g/mol) 190.665
MDL Number MFCD00053211
InChI Key KBAZUXSLKGQRJF-UHFFFAOYSA-N
Synonym trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
PubChem CID 73866
IUPAC Name chloro-dimethyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](C)(CCC(F)(F)F)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369305
View Documents
Thermo Scientific Alfa Aesar
L16666.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11379305
View Documents
Thermo Scientific Alfa Aesar
L16666.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11389305
View Documents
Thermo Scientific Alfa Aesar
L16666.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1481-41-0
190.665
KBAZUXSLKGQRJF-UHFFFAOYSA-N
73866
C[Si](C)(CCC(F)(F)F)Cl
C5H10ClF3Si
MFCD00053211
trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
chloro-dimethyl-(3,3,3-trifluoropropyl)silane

Specifications

118°C to 120°C
C5H10ClF3Si
UN2985
20°C (68°F)
KBAZUXSLKGQRJF-UHFFFAOYSA-N
chloro-dimethyl-(3,3,3-trifluoropropyl)silane
73866
96%
1481-41-0
MFCD00053211
1741726
trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
C[Si](C)(CCC(F)(F)F)Cl
190.665
190.67
Chlorodimethyl-3,3,3-trifluoropropylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications