missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroiodomethane, 98%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent

Specifications
Specifications
Color | Pink-Red to Yellow |
Density | 2.4200g/mL |
Boiling Point | 108.0°C to 109.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | ClCH2I |
Refractive Index | 1.5812 to 1.5832 |
Quantity | 500 mL |
Beilstein | 01,71 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chloroiodomethane, 98%, stabilized, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction