missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Chlorophenol Red Product Code.: 11417254

Thermo Scientific Chemicals Chlorophenol Red

Product Code. 11417254
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11417254

Brand: Thermo Scientific Chemicals B21623.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.

Solubility
Soluble in water (30 mg/ml), ethanol (80 mg/ml), and methyl cellusolve (50 mg/ml).

Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-20-0
Molecular Formula C19H12Cl2O5S
Molecular Weight (g/mol) 423.26
MDL Number MFCD00005877
InChI Key WWAABJGNHFGXSJ-UHFFFAOYSA-N
Synonym chlorophenol red, chlorphenol red, chlorphenolsulfonphthalein, dichlorophenolsulfonephthalein, 3,3-bis 3-chloro-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, chlorophenol red indicator, 3',3-dichlorophenolsulfonphthalein, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-chloro, 3',3'-dichlorophenolsulfonaphthalein, 3',3-dichlorophenol-sulfonphthaleinShow MoreShow Less
PubChem CID 20486
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1

Specifications

Quantity 25 g
Beilstein 354053
Solubility Information Soluble in water (30mg/ml),ethanol (80mg/ml),and methyl cellusolve (50mg/ml).
Formula Weight 432.28
Chemical Name or Material Chlorophenol Red

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10222089 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberB21623.06,B21623.14
Lot Number10151230 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21623.06,B21623.14
Lot Number10168285 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21623.06,B21623.14
Lot NumberF8594B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21623.06,B21623.14
Lot Number10222089 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21623.06,B21623.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Chlorophenol Red >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.