missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline kinase alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13520-1-AP-150UL
This item is not returnable.
View return policy
Description
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.Specifications
Choline kinase alpha | |
Polyclonal | |
Unconjugated | |
CHKA | |
CHETK alpha, CHK, CHKA, Choline kinase alpha, CK, CKI, CK-alpha, EK, Ethanolamine kinase; | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1119, 12660, 29194 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54804, P35790, Q01134 | |
Chka | |
Choline kinase alpha Fusion Protein Ag4761 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |