missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline kinase alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | Choline kinase alpha |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881774
|
Proteintech
13520-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871774
|
Proteintech
13520-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.Specifications
Choline kinase alpha | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O54804, P35790, Q01134 | |
1119, 12660, 29194 | |
Choline kinase alpha Fusion Protein Ag4761 | |
Primary | |
-20°C | |
CHKA |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CHETK alpha, CHK, CHKA, Choline kinase alpha, CK, CKI, CK-alpha, EK, Ethanolamine kinase; | |
Chka | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |