Learn More
Choline kinase alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13520-1-AP-20UL
 View more versions of this product
Product Code. 16881774
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.
Specifications
Specifications
Choline kinase alpha
Polyclonal
Unconjugated
CHKA
CHETK alpha, CHK, CHKA, Choline kinase alpha, CK, CKI, CK-alpha, EK, Ethanolamine kinase;
Rabbit
Antigen Affinity Chromatography
RUO
1119, 12660, 29194
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54804, P35790, Q01134
Chka
Choline kinase alpha Fusion Protein Ag4761
20 μL
Primary
Mouse, Rat, Human