Learn More
Azpack™ Freeze Drying and Injection Vials
Ensure optimum protection of the contents against premature aging. Azpack™ Freeze Drying and Injection Vials protect content integrity and loss of effectiveness.
Brand:  Azpack™ 10653205
 View more versions of this product
Product Code. 10653205
  • 539.00 € / Pack of 441
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from FIOLAX™ neutral clear glass
  • Outstanding chemical resistance, neutrality and impermeability
  • Crimp neck
  • Supplied without caps
Specifications
Specifications
Vial
Clear
Crimp
13mm
6.35 mm
2 mL
FIOLAX™ Glass
441/Pk.
14.25 mm (Neck O.D.)
39.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products