missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromacol 12mm and 13mm Closures for Screw-Thread Vials

Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.

Brand:  Thermo Scientific™ 13-SC-8RT1

179.72 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.


 View more versions of this product

Product Code. 10004012

  • 218.37 € / Pack of 500
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

13mm Screw Cap Prefitted Type 8 Rubber/PTFE Liner for 3.5-HRSV
Black
Chromacol
Rubber/PTFE
Rubber
Screw Thread Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.