missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromacol 12mm and 13mm Closures for Screw-Thread Vials

Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.

55.96 € - 218.37 €

Specifications

Product Line Chromacol
For Use With (Equipment) Screw Thread Vials
View More Specs

Products 3
Product Code Brand ClosureMaterial ClosureColor Price Quantity & Availability  
Product Code Brand ClosureMaterial ClosureColor Price Quantity & Availability  
10004012
View Documents
Thermo Scientific™
13-SC-8RT1
218.37 €
Pack of 500
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
11533710
View Documents
Thermo Scientific™
12-SCWG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11583700
View Documents
Thermo Scientific™
12-SC(R)
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific™ Chromacol Vial Closures are offered with a selection of liner materials and colors.

Specifications

Specifications

Chromacol
Screw Thread Vials
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Chromacol 12mm and 13mm Closures for Screw-Thread Vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.