Learn More
Macherey-Nagel™ Chromafix™ SA Cartridge
180.00 € - 279.00 €
Specifications
Base Material Silica
For Use With (Application) Amino Acids, Amines, Chlorophyll, PCBs
Particle Size 45 μm
Sorbent SA
Type Chromafix SA Cartridge
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10135091
View Documents
Macherey-Nagel™
731832
Adsorbent Weight: 450 mg
210.00 €
Pack of 50
In Stock
to see stock.
Add to basket
10175041
View Documents
Macherey-Nagel™
731831
Adsorbent Weight: 220 mg
180.00 €
Pack of 50
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
10145091
View Documents
Macherey-Nagel™
731833
Adsorbent Weight: 920 mg
279.00 €
Pack of 50
In Stock
to see stock.
Add to basket
Specifications
Silica
45 μm
Chromafix SA Cartridge
50 Pack
Amino Acids, Amines, Chlorophyll, PCBs
SA
60 Å
500 m2/g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ SA Cartridge