Learn More
Pro-Lab Diagnostics™ Nichrome Inoculation Loops
Calibrated to remove a consistent, precise amount of material. Pro-Lab Diagnostics™ Nichrome Inoculation Loops are economical nickel-chrome wire loops in a range of sizes from 1μL to 10μL. The standard and heavy gauge wire loops can be used with insulated holders (sold separately).
44.00 € - 103.00 €
Product Code | Brand | Diameter (Metric) Inside | Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Gauge | Price | Quantity & Availability | |||||
11909054
|
Pro-Lab Diagnostics™
PL.104 |
2.26 mm | Standard |
53.00 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||||
11919054
|
Pro-Lab Diagnostics™
PL.105 |
1.59 mm | Standard |
52.00 €
Pack of 25 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
11999044
|
Pro-Lab Diagnostics™
PL.102 |
2.91 mm | Standard |
103.00 €
Pack of 25 |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
12840155
|
Pro-Lab Diagnostics™
PL.130 |
5.05 mm | Heavy |
44.00 €
Pack of 25 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
12850155
|
Pro-Lab Diagnostics™
PL.131 |
3.57 mm | Heavy |
45.00 €
Pack of 25 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
12830155
|
Pro-Lab Diagnostics™
PL.103 |
2.52 mm | Standard |
52.00 €
Pack of 25 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
12860155
|
Pro-Lab Diagnostics™
PL.133 |
2.52 mm | Heavy |
44.00 €
Pack of 25 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
Description
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)
- Constructed of economical nickel-chrome in standard or heavy gauge wire
- Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
- Packaged in sets of 25 loops
- Holders with insulated handles sold separately
Specifications
Chrome-Nickel | |
Inoculating Loop |