missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Potassium Sulfate Dodecahydrate, 98.0 to 102.0%, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥98%
28.92 € - 115.44 €
Chemical Identifiers
CAS | 7788-99-0 |
---|---|
Molecular Formula | CrH24KO20S2 |
Molecular Weight (g/mol) | 499.386 |
MDL Number | MFCD00149917 |
InChI Key | ZFVHBEKVAITXHW-UHFFFAOYSA-J |
Synonym | chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p |
PubChem CID | 24596 |
IUPAC Name | potassium;chromium(3+);disulfate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7788-99-0 | |
499.386 | |
ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
24596 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] |
CrH24KO20S2 | |
MFCD00149917 | |
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
potassium;chromium(3+);disulfate;dodecahydrate |
Specifications
89°C | |
Violet | |
CrK(SO4)2 · 12H2O | |
NONH for all modes of transport | |
ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
potassium;chromium(3+);disulfate;dodecahydrate | |
24596 | |
98.0 to 102.0% | |
1.8320g/cm3 |
7788-99-0 | |
CrH24KO20S2 | |
MFCD00149917 | |
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] | |
499.386 | |
499.4g/mol | |
ACS Reagent | |
Chromium(Iii) Potassium Sulfate Dodecahydrate |
Safety and Handling
missing translation for 'einecsNumber' : 233-401-6
missing translation for 'rtecsNumber' : GB6850000