Learn More
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Chromium, Cr, CAS Number-7440-47-3, chrome, chromium, elemental, ccris 159, chrome french, compounds, chrom, chrom german, cromo, metal, chromium, metal, 500g, Gray powder, 99.95% (Metals basis), CHEBI:28073, DOT Class: 4.1, Packaging Group: III, 231-157-5, H228
114.00 € - 1212.00 €
Chemical Identifiers
CAS | 7440-47-3 |
---|---|
Molecular Formula | Cr |
Molecular Weight (g/mol) | 52.00 |
MDL Number | MFCD00010944 |
InChI Key | VYZAMTAEIAYCRO-UHFFFAOYSA-N |
Synonym | chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 |
PubChem CID | 23976 |
ChEBI | CHEBI:28073 |
IUPAC Name | chromium |
SMILES | [Cr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381329
|
Thermo Scientific Alfa Aesar
013796.14 |
25 g |
114.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371329
|
Thermo Scientific Alfa Aesar
013796.22 |
100 g |
318.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391329
|
Thermo Scientific Alfa Aesar
013796.36 |
500 g |
1212.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-47-3 | |
52.00 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
23976 | |
chromium |
Cr | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
CHEBI:28073 | |
[Cr] |
Specifications
Silver | |
Cr | |
UN3089 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
chromium | |
23976 | |
99.95% | |
Odorless | |
Chromium powder |
7440-47-3 | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
[Cr] | |
52.00 | |
CHEBI:28073 | |
−200 mesh | |
(metals basis) |
Safety and Handling
GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 231-157-5
missing translation for 'rtecsNumber' : GB4200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only