missing translation for 'onlineSavingsMsg'
Learn More

Chromium silicon monoxide, 99.9% (metals basis), Thermo Scientific™

49.00 € - 176.00 €

Chemical Identifiers

CAS 37264-20-3
Molecular Formula CrOSi
Molecular Weight (g/mol) 96.08
MDL Number MFCD02091702
InChI Key ASAMIKIYIFIKFS-UHFFFAOYSA-N
Synonym chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
PubChem CID 57464841
IUPAC Name chromium;oxoniumylidynesilanide
SMILES [O+]#[Si-].[Cr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313827
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035667.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11323827
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035667.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

37264-20-3
96.08
ASAMIKIYIFIKFS-UHFFFAOYSA-N
57464841
[O+]#[Si-].[Cr]
CrOSi
MFCD02091702
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
chromium;oxoniumylidynesilanide

Specifications

37264-20-3
MFCD02091702
It is insoluble in water.
[O+]#[Si-].[Cr]
96.08
99.9%
Chromium silicon monoxide
CrOSi
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
ASAMIKIYIFIKFS-UHFFFAOYSA-N
chromium;oxoniumylidynesilanide
57464841
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications