Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins.
Brand:  Proteintech nta-200
 View more versions of this product
Product Code. 17333393
  • 3237.00 € / 200 reactions
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mNeonGreen, Branchiostoma lanceolatum
Alpaca
IP, CoIP, ChIP, RIP
Agarose
200 Reactions