missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins.
238.00 € - 5899.00 €
Specifications
Content And Storage | Upon receipt store at 4°C. Do not freeze. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | mNeonGreen, Branchiostoma lanceolatum |
Label or Dye | Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303393
|
Proteintech
nta-10 |
10 Reactions |
238.00 €
10 reactions |
In Stock
Log in to see stock. |
|||||
17313393
|
Proteintech
NTA-100 |
100 Reactions |
1798.00 €
100 reactions |
In Stock
Log in to see stock. |
|||||
17323393
|
Proteintech
nta-20 |
20 Reactions |
397.00 €
20 reactions |
In Stock
Log in to see stock. |
|||||
17333393
|
Proteintech
nta-200 |
200 Reactions |
3237.00 €
200 reactions |
In Stock
Log in to see stock. |
|||||
17343393
|
Proteintech
NTA-400 |
400 Reactions |
5899.00 €
400 reactions |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mNeonGreen, Branchiostoma lanceolatum | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose |