missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human Ig, Lambda-LC-specific VHH, Biotinylated
Nano-CaptureLigand for human Ig: lambda-specific single domain antibody/VHH [CTK0110], biotinylated.
Brand: Proteintech SHULB-1-10
This item is not returnable.
View return policy
Description
- The Nano-CaptureLigand™ human Ig, lambda-LC captures and immobilizes human Ig lambda light chain (LC) antibodies to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
- Consists of a biotinylated Nanobody that specifically binds to human Ig lambda LC antibodies with high affinity and in a site-directed manner
Specifications
10 μL | |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
Lambda-LC of Human Ig lambda Antibodies | |
Monoclonal | |
BLI, SPR, ELISA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech ChromoTek Nano-CaptureLigand™ Human Ig, Lambda-LC-specific VHH, Biotinylated > 10 μL
Spot an opportunity for improvement?Share a Content Correction