Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
PARP1-Trap Agarose is an affinity reagent for IP of human PARP1.
Brand: Proteintech xta-10
This item is not returnable.
View return policy
Description
- Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
Human PARP1, does not bind to PARP2, 3, and 9 | |
Alpaca |
IP, CoIP | |
Agarose | |
10 Reactions |