missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
PARP1-Trap Agarose is an affinity reagent for IP of human PARP1.
238.00 € - 5899.00 €
Specifications
Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
---|---|
For Use With (Application) | IP, CoIP |
Target | Human PARP1, does not bind to PARP2, 3, and 9 |
Label or Dye | Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17353563
|
Proteintech
xta-10 |
10 Reactions |
238.00 €
10 reactions |
In Stock
Log in to see stock. |
|||||
17363563
|
Proteintech
xta-100 |
100 Reactions |
1798.00 €
100 reactions |
In Stock
Log in to see stock. |
|||||
17373563
|
Proteintech
xta-20 |
20 Reactions |
397.00 €
20 reactions |
In Stock
Log in to see stock. |
|||||
17383563
|
Proteintech
xta-200 |
200 Reactions |
3237.00 €
200 reactions |
In Stock
Log in to see stock. |
|||||
17393563
|
Proteintech
xta-400 |
400 Reactions |
5899.00 €
400 reactions |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
Human PARP1, does not bind to PARP2, 3, and 9 | |
Alpaca |
IP, CoIP | |
Agarose |