Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
PARP1-Trap Agarose is an affinity reagent for IP of human PARP1.
Brand:  Proteintech xta-200
 View more versions of this product
Product Code. 17383563
  • 3237.00 € / 200 reactions
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
Human PARP1, does not bind to PARP2, 3, and 9
Alpaca
IP, CoIP
Agarose
200 Reactions