Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads
PARP1-Trap Agarose is an affinity reagent for IP of human PARP1.
Brand:  Proteintech xta-400
 View more versions of this product
Product Code. 17393563
  • 5899.00 € / 400 reactions
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifications
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
Human PARP1, does not bind to PARP2, 3, and 9
Alpaca
IP, CoIP
Agarose
400 Reactions