missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | CHTF18 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16821044
|
Proteintech
13030-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16811044
|
Proteintech
13030-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHTF18, CHTF8.Specifications
CHTF18 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8BIW9, Q8WVB6 | |
214901, 287146, 63922 | |
CHTF18 Fusion Protein Ag3672 | |
Primary | |
-20°C | |
CHTF18 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, CHTF18, Ctf18, hCTF18, RUVBL | |
Chtf18 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |