Learn More
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13030-1-AP-150UL
 View more versions of this product
Product Code. 16811044
  • 410.00 € / 150µL
Explore more special offers
Description
Description
CHTF18, CHTF8.
Specifications
Specifications
CHTF18
Polyclonal
Unconjugated
CHTF18
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, CHTF18, Ctf18, hCTF18, RUVBL
Rabbit
Antigen Affinity Chromatography
RUO
214901, 287146, 63922
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BIW9, Q8WVB6
Chtf18
CHTF18 Fusion Protein Ag3672
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated