Learn More
Tocris Bioscience™ Cinacalcet hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand:  Tocris Bioscience™ 6170/10
114.03 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18734739
  • 145.00 € / 10mg
Explore more special offers
Description
Description
Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.
Specifications
Specifications
Inhibitor
364782-34-3
393.87g/mol
Calcium-Sensing Receptor Agonists
Calcium-Sensing Receptor
C22H22F3N.HCl
Store at -20°C