missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cinacalcet hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
145.00 € - 524.00 €
Specifications
Product Type | Inhibitor |
---|---|
Target Proteins | Calcium-Sensing Receptor |
CAS | 364782-34-3 |
Molecular Formula | C22H22F3N.HCl |
Formula Weight | 393.87g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734739
|
Tocris Bioscience™
6170/10 |
10 mg |
145.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736258
|
Tocris Bioscience™
6170/50 |
50 mg |
524.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.Specifications
Inhibitor | |
364782-34-3 | |
393.87g/mol | |
Calcium-Sensing Receptor Agonists |
Calcium-Sensing Receptor | |
C22H22F3N.HCl | |
Store at -20°C |
For Research Use Only