Learn More
Thermo Scientific Chemicals Ciprofloxacin, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Ciprofloxacin is considered a synthetic broad-spectrum second-generation compound with antibiotic activity mainly against gram-negative bacteria
• It inhibits bacterial DNA gyrase and topoisomerase IV, enzymes that are essential for DNA replication, cell division, and causes double-strand breaks in the bacterial chromosome
Applications
• Ciprofloxacin presents more activity against gram-negative bacteria than gram-positive bacteria.
• Ciprofloxacin does not present cross-resistance between fluoroquinolones and other antibiotics classes, which is beneficial in laboratory experiments when other antibiotics are no longer effective

Specifications
Specifications
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 5 g |
Merck Index | 15,2313 |
Solubility Information | Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in hot dmso,soluble in hot methanol |
Formula Weight | 331.34 |
Percent Purity | 98% |
Chemical Name or Material | Ciprofloxacin |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461570 | Certificate of Analysis | 09/04/2024 | 449620250,449620050 |
A0452189 | Certificate of Analysis | 02/05/2023 | 449620250,449620050 |
A0392198 | Certificate of Analysis | 14/12/2022 | 449620250,449620050 |
A0413292 | Certificate of Analysis | 13/12/2022 | 449620250,449620050 |
A0410522 | Certificate of Analysis | 13/12/2022 | 449620250,449620050 |
Safety and Handling
- Ciprofloxacin
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.