Learn More
cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
49.00 € - 118.30 €
Chemical Identifiers
CAS | 156-59-2 |
---|---|
Molecular Formula | C2H2Cl2 |
Molecular Weight (g/mol) | 96.94 |
MDL Number | MFCD00000925 |
InChI Key | KFUSEUYYWQURPO-UPHRSURJSA-N |
Synonym | cis-1,2-dichloroethene, z-1,2-dichloroethene, cis-1,2-dichloroethylene, z-1,2-dichloroethylene, cis-dichloroethylene, acetalyne dichloride, acetylene dichloride, cis, ethene, 1,2-dichloro-, z, cis-acetylene dichloride, unii-fyo9g15jyd |
PubChem CID | 643833 |
ChEBI | CHEBI:28805 |
IUPAC Name | (Z)-1,2-dichloroethene |
SMILES | C(=CCl)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-59-2 | |
96.94 | |
KFUSEUYYWQURPO-UPHRSURJSA-N | |
643833 | |
(Z)-1,2-dichloroethene |
C2H2Cl2 | |
MFCD00000925 | |
cis-1,2-dichloroethene, z-1,2-dichloroethene, cis-1,2-dichloroethylene, z-1,2-dichloroethylene, cis-dichloroethylene, acetalyne dichloride, acetylene dichloride, cis, ethene, 1,2-dichloro-, z, cis-acetylene dichloride, unii-fyo9g15jyd | |
CHEBI:28805 | |
C(=CCl)Cl |
Specifications
156-59-2 | |
Colorless | |
60.0°C | |
Authentic | |
C2H2Cl2 | |
1.4475 to 1.4495 | |
01,188 | |
15,87 | |
cis-1,2-dichloroethene, z-1,2-dichloroethene, cis-1,2-dichloroethylene, z-1,2-dichloroethylene, cis-dichloroethylene, acetalyne dichloride, acetylene dichloride, cis, ethene, 1,2-dichloro-, z, cis-acetylene dichloride, unii-fyo9g15jyd | |
C(=CCl)Cl | |
0.003% MEHQ | |
643833 | |
96.94 | |
Liquid |
-80.0°C | |
1.2800g/mL | |
6°C | |
96% min. (GC) | |
ClCH=CHCl | |
MFCD00000925 | |
13,293 | |
1.28 | |
KFUSEUYYWQURPO-UPHRSURJSA-N | |
(Z)-1,2-dichloroethene | |
96.94 | |
CHEBI:28805 | |
97% | |
cis-1, 2-Dichloroethylene, 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-859-7
missing translation for 'rtecsNumber' : KV9420000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.