Learn More
Thermo Scientific Chemicals cis-5,8,11,14,17-Eicosapentaenoic acid
291.45 € - 556.77 €
Chemical Identifiers
CAS | 10417-94-4 |
---|---|
Molecular Formula | C20H30O2 |
Synonym | EPA; Timnodonic acid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497269
|
Thermo Scientific Chemicals
J66060.MC |
100 mg |
291.45 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407279
|
Thermo Scientific Chemicals
J66060.ME |
500 mg |
556.77 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10417-94-4 | |
C20H30O2 | |
EPA; Timnodonic acid |
Specifications
Colorless | |
10417-94-4 | |
302.45 | |
93°C (199°F) | |
Air and Light Sensitive | |
Soluble in methanol | |
Oil | |
JAZBEHYOTPTENJ-JLNKQSITSA-N | |
(5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid |
-30°C to -10°C | |
C20H30O2 | |
EPA; Timnodonic acid | |
0.943 | |
-54° | |
cis-5,8,11,14,17-Eicosapentaenoic acid | |
1714433 | |
CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O | |
302.46 |
Safety and Handling
Recommended Storage : Store at -20°C; Store under Argon
- cis-5,8,11,14,17-Eicosapentaenoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Eicosapentaenoic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.