missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific Chemicals
40.70 € - 151.00 €
Chemical Identifiers
CAS | 60662-54-6 |
---|---|
Molecular Formula | C6H12O3·2H2O |
Molecular Weight (g/mol) | 168.19 |
InChI Key | MAPLMYMZLRIJSP-UHFFFAOYSA-N |
Synonym | cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate |
PubChem CID | 16211857 |
IUPAC Name | cyclohexane-1,3,5-triol;dihydrate |
SMILES | C1C(CC(CC1O)O)O.O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15590594
|
Thermo Scientific Acros
458950010 |
1 g |
40.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15500604
|
Thermo Scientific Acros
458950050 |
5 g |
151.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60662-54-6 | |
168.19 | |
cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate | |
cyclohexane-1,3,5-triol;dihydrate |
C6H12O3·2H2O | |
MAPLMYMZLRIJSP-UHFFFAOYSA-N | |
16211857 | |
C1C(CC(CC1O)O)O.O.O |
Specifications
60662-54-6 | |
Authentic | |
Glass Bottle | |
cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate | |
MAPLMYMZLRIJSP-UHFFFAOYSA-N | |
cyclohexane-1,3,5-triol;dihydrate | |
16211857 | |
98% |
108.0°C to 110.0°C | |
97.5% min. (GC) | |
C6H12O3·2H2O | |
Solubility in water: soluble in hot water. | |
C1C(CC(CC1O)O)O.O.O | |
168.19 | |
168.19 | |
cis,cis-1,3,5-Cyclohexanetriol dihydrate |
RUO – Research Use Only