Learn More
Citicoline, MedChemExpress
MedChemExpress Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.

70.20 € - 125.55 €
Specifications
Chemical Name or Material | Citicoline |
---|---|
CAS | 987-78-0 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C14H26N4O11P2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478436
![]() |
Medchem Express
HY-B0739 |
100 mg |
70.20 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16472206
![]() |
Medchem Express
HY-B0739 |
500 mg |
125.55 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.Specifications
Citicoline | |
White | |
C14H26N4O11P2 | |
H2O : 50 mg/mL (102.39 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.05 mM; ultrasonic and warming and heat to 80°C) | |
488.32 | |
99.51% | |
Solid |
987-78-0 | |
Research | |
Cytidine diphosphate-choline CDP-Choline Cytidine 5'-diphosphocholine | |
O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)N)=O)O2)O)[C@H]2COP(OCC[N+](C)(C)C)(OP([O-])(O)=O)=O | |
488.32 | |
Research | |
Neuroscience-Neurodegeneration |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.