Learn More
Citicoline, MedChemExpress
MedChemExpress Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.
Brand: Medchem Express HY-B0739

Description
Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.Specifications
Citicoline | |
White | |
C14H26N4O11P2 | |
Cytidine diphosphate-choline CDP-Choline Cytidine 5'-diphosphocholine | |
O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)N)=O)O2)O)[C@H]2COP(OCC[N+](C)(C)C)(OP([O-])(O)=O)=O | |
488.32 | |
Research | |
Neuroscience-Neurodegeneration |
987-78-0 | |
Research | |
500 mg | |
H2O : 50 mg/mL (102.39 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.05 mM; ultrasonic and warming and heat to 80°C) | |
488.32 | |
99.51% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only