missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™

C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid

39.20 € - 330.00 €

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
MDL Number 149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10345410
View accessory products View Documents Promotion Details
Fisher Chemical
C/6200/53
500 g Plastic powder jar
58.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
10655102
View Documents Promotion Details
Fisher Chemical
C/6200/60
1 kg Plastic powder jar
39.20 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10796281
View Documents Promotion Details
Fisher Chemical
C/6200/62
2.5 kg Plastic powder jar
187.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10665102
View Documents Promotion Details
Fisher Chemical
C/6200/65
5 kg Plastic keg
330.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid

Specifications

Chemical Identifiers

5949-29-1
210.14
YASYEJJMZJALEJ-UHFFFAOYSA-N
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
C6H10O8
149972
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate

Specifications

1.8
135°C
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
210.15g/mol
5949-29-1
White
149972
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
210.14
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Citric acid monohydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash A§ thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications