Learn More
Hirschmann™ Class A Measuring Cylinders, Amber Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Amber Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2210160
 View more versions of this product
Product Code. 13117939
  • 33.15 € / Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With spout, main point ring amber stain graduation
  • Conformity certified, with dated batch identification
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
DIN EN ISO 4788
140 mm
10 mL
0.01
A
Glass
0.2 mL