missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Class AS BLAUBRAND™ Graduated Pipette

Class AS BLAUBRAND™ Graduated Pipette is type 3 and features zero on top.

75.05 € - 100.00 €

Specifications

Accuracy ±0.01 mL
Certifications/Compliance DIN EN ISO 835
Class AS
Description TD, Ex
Material AR-GLAS
View More Specs

Products 2
Product Code Brand Graduations Price Quantity & Availability  
Product Code Brand Graduations Price Quantity & Availability  
10657561
View Documents
BRAND™
27309
0.02 mL
100.00 €
Pack of 12
In Stock
to see stock.
Add to basket
 
10473531
View Documents
BRAND™
27710
0.1 mL
75.05 €
Pack of 12
In Stock
to see stock.
Add to basket
 
Description

Description

  • AR-GLAS™
  • Type 3 (zero on top)
  • Calibrated to deliver (TD, Ex)
  • Waiting time 5 seconds
  • Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
Specifications

Specifications

±0.01 mL
AS
AR-GLAS
DIN EN ISO 835
TD, Ex
2 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Class AS BLAUBRAND™ Graduated Pipette

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.