Learn More
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.
Brand:  Hirschmann™ 7501194
 View more versions of this product
Product Code. 11671409
  • 142.00 € / Pack of 3
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Blue graduated
  • Main point ring graduation
  • Hexagonal base
Specifications
Specifications
B
Round
Blue
440 mm
Reusable
Precise Liquid Measurement and Fluid Transfer in the Laboratory
Hexagonal
Graduated Measuring Cylinder