Learn More
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.
Brand: Hirschmann™ 7501186
This item is not returnable.
View return policy
Description
- Acc. to DIN 12681/ISO 6706
- PP
- Raised scale
- Blue graduated
- Main point ring graduation
- Hexagonal base
Specifications
B | |
Round | |
Blue | |
315 mm |
Reusable | |
Precise Liquid Measurement and Fluid Transfer in the Laboratory | |
Hexagonal | |
Graduated Measuring Cylinder |