Learn More
Prestige Medical™ Classic Autoclave
Brand:  Prestige Medical™ 210004UK
 View more versions of this product
Product Code. 12300479
  • 2835.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The availability of a wide range of model sizes, features and internal furniture options make the Prestige Medical Classic a popular choice where a cost effective, portable autoclave is needed. Fully automatic, simply press the button to start. Lightweight and portable, ideal for a variety of sterilisation solutions.
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Pressure and temperature gauges
  • Complete with instrument basket
  • CE marked
  • Comprehensive one year warranty (UK)
  • Automatic electronic time and temperature control with end of cycle cut off and buzzer
  • Indicator lights for power on, heating, sterilisation cycle, fault condition and sterilisation complete
  • Incorrectly fitted lid does not allow pressure to build up
  • Steam release valve automatically releases excess pressure
  • Safety fuse operates if temperature is too high
  • If water level is too low or sterilisation cycle is interrupted a red fault lights operates and the autoclave switches off
  • DoH performance validated model with thermocouple entry port, pressure and temperature gauge For the sterilisation of unwrapped solid instruments and devices.
Specifications
Specifications
12 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Prestige Medical™ Classic Autoclave > Capacity: 12L