Learn More
Prestige Medical™ Classic Autoclave
1435.00 € - 2835.00 €
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12300479
View accessory products View Documents
Prestige Medical™
210004UK
12 L
2835.00 €
Each
In Stock
to see stock.
Add to basket
12755345
View accessory products View Documents
Prestige Medical™
210001UK
9 L
1435.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
Description
Description
The availability of a wide range of model sizes, features and internal furniture options make the Prestige Medical Classic a popular choice where a cost effective, portable autoclave is needed. Fully automatic, simply press the button to start. Lightweight and portable, ideal for a variety of sterilisation solutions.
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Pressure and temperature gauges
  • Complete with instrument basket
  • CE marked
  • Comprehensive one year warranty (UK)
  • Automatic electronic time and temperature control with end of cycle cut off and buzzer
  • Indicator lights for power on, heating, sterilisation cycle, fault condition and sterilisation complete
  • Incorrectly fitted lid does not allow pressure to build up
  • Steam release valve automatically releases excess pressure
  • Safety fuse operates if temperature is too high
  • If water level is too low or sterilisation cycle is interrupted a red fault lights operates and the autoclave switches off
  • DoH performance validated model with thermocouple entry port, pressure and temperature gauge For the sterilisation of unwrapped solid instruments and devices.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Prestige Medical™ Classic Autoclave