missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva CleanGel for IEF
Perform sensitive procedures using precast gels that are carefully washed and dried after casting to remove catalysts and unpolymerized material
Brand: Cytiva 18103532
This item is not returnable.
View return policy
Description
- Customize IEF separations by reswelling in a suitable mixture of carrier ampholytes and other additives such as detergents or denaturants
- Handle mechanically stable, plastic-backed gels with confidence
- Offer improved gel performance with faster and more stable gradients, fewer artifacts and easier zymogram assays
- Before use, rehydrate using the convenient GelPool accessory (Mfr. No. 18-1031-58); swelling returns the gel to its original thickness of 0.5mm
- Gel composition: Polyacrylamide gel (5%T/3%C)

Specifications
CleanGel IEF | |
Precast Gel | |
Analytical chemistry, Research CleanGel IEF gels offer improved gel performance with faster and more stable gradients, fewer artifacts and easier zymogram assays | |
5 | |
0.019 in. | |
110 mm |
Clear | |
Isoelectric Focusing | |
Polyester film | |
-20°C | |
0.05 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva CleanGel for IEF > CleanGel IEF
Spot an opportunity for improvement?Share a Content Correction