missing translation for 'onlineSavingsMsg'
Learn More

Purus™ Pure Comfort Cleanroom Socks

Purus™ Cleanroom Socks are designed for cleanroom use and comfort.

1595.00 €

Specifications

Color White
For Use With (Application) Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors
Material Polyester
Sterility Non-sterile
Cleanroom Class Class 10-1000 (ISO 4-6)
View More Specs

Products 4
Product Code Brand Disposable Size Price Quantity & Availability  
Product Code Brand Disposable Size Price Quantity & Availability  
17793799
View Documents
Purus™
CRS-L-S
Reusable Small
1595.00 €
200 pairs
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
17783799
View Documents
Purus™
CRS-L-M
Reusable Medium
1595.00 €
200 pairs
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
17773799
View Documents
Purus™
CRS-L-L
Reusable Large
1595.00 €
200 pairs
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
17703809
View Documents
Purus™
CRS-L-XL
Reusable X-Large
1595.00 €
200 pairs
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
Description

Description

  • Available in both disposable and launderable styles
  • Made from 100% polyester
  • Cleanroom compatible
  • Comfortable for extended use
  • No particle generation
Specifications

Specifications

White
Polyester
Class 10-1000 (ISO 4-6)
Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.