Learn More
CleanSpace™ HALO Half Mask for PAPR Assembly
Specifically designed to work to complete the CleanSpace HALO system and clip quickly onto the power units. Half masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
Brand:  CleanSpace™ CS3005
 View more versions of this product
Product Code. 15910042
  • 199.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
  • Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Mask Fit Test adaptor available for TSI Portacount™ fit testing
  • Easy removal
  • Compatible with most cleaning and disinfection protocols
  • Transparent and facilitates communication
Specifications
Specifications
AS/NZS1716: 2012
PAPR Half Mask
Half mask only
No
EN 12942
Large
Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders
CS3005
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.