Learn More
CleanSpace™ HALO Half Mask for PAPR Assembly
Specifically designed to work to complete the CleanSpace HALO system and clip quickly onto the power units. Half masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
199.00 €
Specifications
Certifications/Compliance AS/NZS1716: 2012
Certifications/Compliance (Europe) EN 12942
Product Type PAPR Half Mask
Facepiece/Headtop Model Half mask only
For Use With (Application) Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15900042
View Documents
CleanSpace™
CS3004
Medium
199.00 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15910042
View Documents
CleanSpace™
CS3005
Large
199.00 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
  • Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Mask Fit Test adaptor available for TSI Portacount™ fit testing
  • Easy removal
  • Compatible with most cleaning and disinfection protocols
  • Transparent and facilitates communication
Specifications
Specifications
AS/NZS1716: 2012
PAPR Half Mask
Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders
EN 12942
Half mask only
No
Videos
SDS
Documents
Documents
Product Certifications
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.